Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem198b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178066.2 | NP_835167.1 | 3 | 433 | ||
GenBank mRNA | AK054166.1 | - | 2 | 890 | ||
BC037624.1 | - | 3 | 433 | 4507848 |