Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178930.3 | NP_849261.2 | 3 | 397 | ||
GenBank mRNA | AK039570.1 | - | 3 | 415 | ||
AK041959.1 | - | 3 | 427 | |||
AK143358.1 | - | 3 | 409 | |||
BC011335.1 | - | 3 | 361 | |||
BC076569.1 | - | 3 | 397 | 6409170 | ||
BC082336.1 | - | 3 | 392 |