Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A130010J15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160359.1 | NP_001153831.1 | 2 | 525 | ||
NM_001160360.1 | NP_001153832.1 | 2 | 373 | |||
NM_181048.3 | NP_851391.1 | 2 | 535 | |||
GenBank mRNA | AK037364.1 | - | 2 | 532 | ||
AK047686.1 | - | 2 | 469 | |||
AK077339.1 | - | 2 | 374 | |||
AK084448.1 | - | 2 | 421 | |||
BC055955.1 | - | 2 | 470 | 4527064 |