Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181072.3 | NP_851417.2 | 3, 4 | 591 | ||
GenBank mRNA | AK009155.1 | - | 3, 4 | 592 | ||
AK009427.1 | - | 3, 4 | 592 | |||
AK029145.1 | - | 3, 4 | 593 | |||
BC040786.1 | - | 3, 4 | 579 | |||
BC051391.1 | - | 3, 4 | 573 | 5053090 |