Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079880.1 | NP_001073349.1 | 10, 11 | 1803 | ||
NM_001079881.1 | NP_001073350.1 | 10, 11 | 1667 | |||
NM_001079882.1 | NP_001073351.1 | 8, 9 | 1404 | |||
NM_016457.4 | NP_057541.2 | 9, 10 | 2082 | |||
XM_005258716.2 | XP_005258773.2 | 8, 9 | 1060 | |||
GenBank mRNA | AF309082.1 | - | 9, 10 | 1364 | ||
AK074673.1 | - | 8, 9 | 1043 | |||
AK075266.1 | - | 8, 9 | 1153 | |||
AK092896.1 | - | 9, 10 | 1454 | |||
AK095884.1 | - | 10, 11 | 1346 | |||
AK300339.1 | - | 10, 11 | 1617 | |||
AK303629.1 | - | 8, 9 | 1058 | |||
AL050147.1 | - | 3, 4 | 319 | |||
BC025307.1 | - | 5, 6 | 653 |