Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lingo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181074.4 | NP_851419.1 | 4 | 750 | ||
GenBank mRNA | AF373780.1 | - | 1 | 302 | ||
AK027262.2 | - | 2 | 373 | |||
AK034084.1 | - | 2 | 850 | |||
BC052384.1 | - | 4 | 750 | 5703857 | ||
BC065696.1 | - | 4 | 695 | 5685897 |