Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp6nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181399.3 | NP_852064.2 | 1, 2 | 133 | ||
GenBank mRNA | BC051184.1 | - | 1, 2 | 136 | ||
BQ938063.1 | - | 1, 2 | 133 |