Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181404.5 | NP_852069.4 | 8 | 3650 | ||
GenBank mRNA | AK084638.1 | - | 1 | 100 | ||
AK122214.1 | - | 4 | 2016 | |||
AK147945.1 | - | 5 | 3237 | |||
BC050250.1 | - | 3 | 457 | |||
BC079563.1 | - | 4 | 2977 | 30535512 |