Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181407.4 | NP_852072.2 | 1, 2 | 284 | ||
GenBank mRNA | AK032196.1 | - | 1, 2 | 285 | ||
AK080798.1 | - | 1, 2 | 217 | |||
AK133593.1 | - | 2, 3 | 272 | |||
BC099935.1 | - | 1, 2 | 380 | 30097608 |