Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178930.3 | NP_849261.2 | 5, 6 | 667 | ||
GenBank mRNA | AK039570.1 | - | 5, 6 | 685 | ||
AK041959.1 | - | 5, 6 | 697 | |||
AK143358.1 | - | 5, 6 | 679 | |||
AK172919.1 | - | 2, 3 | 260 | |||
BC011335.1 | - | 5, 6 | 631 | |||
BC076569.1 | - | 5, 6 | 667 | 6409170 | ||
BC082336.1 | - | 5, 6 | 662 |