Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc51b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178933.2 | NP_849264.1 | 4 | 332 | ||
GenBank mRNA | AK002463.1 | - | 4 | 463 | ||
AK004188.1 | - | 4 | 515 | |||
AY279396.1 | - | 4 | 332 | |||
BC115606.1 | - | 3 | 311 | 40102690 | ||
BC115607.1 | - | 3 | 311 | 40102691 |