Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181407.4 | NP_852072.2 | 5, 6 | 806 | ||
GenBank mRNA | AK032196.1 | - | 5, 6 | 807 | ||
AK035780.1 | - | 6, 7 | 2819 | |||
AK080798.1 | - | 5, 6 | 739 | |||
AK133593.1 | - | 6, 7 | 794 | |||
BC099935.1 | - | 5, 6 | 902 | 30097608 |