Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aftph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252503.2 | NP_001239432.1 | 2 | 561 | ||
NM_001290545.1 | NP_001277474.1 | 2 | 561 | |||
NM_181411.4 | NP_852076.1 | 2 | 561 | |||
GenBank mRNA | AK029681.1 | - | 2 | 460 | ||
AK030572.1 | - | 2 | 505 | |||
AK042681.1 | - | 2 | 499 | |||
AK138793.1 | - | 2 | 483 | |||
BC052036.1 | - | 2 | 489 | 5702534 |