Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039194.3 | NP_001034283.1 | 5, 6 | 623 | ||
NM_001284300.1 | NP_001271229.1 | 5, 6 | 623 | |||
NM_153779.2 | NP_722474.2 | 5, 6 | 616 | |||
NM_178058.4 | NP_835159.1 | 5, 6 | 623 | |||
GenBank mRNA | AK017403.1 | - | 5, 6 | 621 | ||
AK085656.1 | - | 5, 6 | 575 | |||
AK147741.1 | - | 5, 6 | 600 | |||
AK155240.1 | - | 5, 6 | 563 | |||
BC038129.1 | - | 5, 6 | 616 | 1395873 |