Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001636.3 | NP_001627.2 | 1480 | |||
GenBank mRNA | BC014775.1 | - | 1433 | 3458777 | ||
BC031912.1 | - | 1380 | 5141625 |