Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178912.3 | NP_849243.2 | 21, 22 | 5707 | ||
GenBank mRNA | AK028941.1 | - | 4, 5 | 1076 | ||
AK162428.1 | - | 2, 3 | 747 | |||
AK173211.1 | - | 21, 22 | 5727 | |||
BC150785.1 | - | 21, 22 | 5701 | 9087696 |