Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kazald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178929.4 | NP_849260.2 | 5 | 1033 | ||
GenBank mRNA | AK030678.1 | - | 5 | 1032 | ||
AK155095.1 | - | 6 | 1278 | |||
BC115642.1 | - | 5 | 1024 | 40103018 | ||
BC115643.1 | - | 5 | 1016 |