Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178930.3 | NP_849261.2 | 7, 8 | 837 | ||
GenBank mRNA | AK039570.1 | - | 7, 8 | 855 | ||
AK041959.1 | - | 7, 8 | 867 | |||
AK143358.1 | - | 7, 8 | 849 | |||
AK172919.1 | - | 4, 5 | 430 | |||
BC004807.1 | - | 2, 3 | 143 | |||
BC011335.1 | - | 7, 8 | 801 | |||
BC076569.1 | - | 7, 8 | 837 | 6409170 | ||
BC082336.1 | - | 7, 8 | 832 |