Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180974.4 | NP_851305.2 | 2, 3 | 732 | ||
GenBank mRNA | AK030735.1 | - | 2, 3 | 687 | ||
AK154401.1 | - | 2, 3 | 724 | |||
BC138824.1 | - | 2, 3 | 619 | |||
BC138825.1 | - | 2, 3 | 619 | |||
CB524750.1 | - | 1, 2 | 126 |