Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610524H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181075.3 | NP_851420.1 | 2 | 322 | ||
GenBank mRNA | AK012147.1 | - | 2 | 311 | ||
AK148078.1 | - | 2 | 297 | |||
BC089033.1 | - | 2 | 302 | |||
BC096574.1 | - | 2 | 313 | 1495259 | ||
BY090347.1 | - | 2 | 322 |
Gene Symbol: 1500011B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161363.1 | - | 2 | 333 | ||
AK165632.1 | - | 2 | 306 | |||
BC061238.1 | - | 2 | 315 |