Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp6nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080548.1 | NP_001074017.1 | 11 | 973 | ||
NM_181399.3 | NP_852064.2 | 10 | 812 | |||
GenBank mRNA | AK084045.1 | - | 11 | 797 | ||
AK129037.1 | - | 11 | 825 | |||
AK133119.1 | - | 2 | 51 | |||
AK155073.1 | - | 11 | 974 | |||
BC051184.1 | - | 10 | 815 | |||
CN455065.1 | - | 3 | 202 |