Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181422.3 | NP_852087.2 | 4 | 830 | ||
GenBank mRNA | AB290927.1 | - | 4 | 662 | ||
AF271381.1 | - | 4 | 796 | |||
AK046772.1 | - | 4 | 831 | |||
BC116297.2 | - | 4 | 761 | 40094997 | ||
BC116323.2 | - | 4 | 761 | 40096335 |