Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007751.3 | NP_031777.1 | 1 | 196 | ||
GenBank mRNA | AA028501.1 | - | 1 | 173 | ||
AA511205.1 | - | 1 | 150 | |||
AK003116.1 | - | 1 | 139 | |||
BC027531.1 | - | 1 | 140 | 1515187 | ||
BC086930.1 | - | 1 | 159 | 6752860 | ||
BE626379.1 | - | 1 | 196 | |||
U15541.1 | - | 1 | 146 |