Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008631.2 | NP_032657.1 | 2 | 124 | ||
GenBank mRNA | AK131894.1 | - | 2 | 124 | ||
BC099850.1 | - | 2 | 83 | |||
BC099851.1 | - | 2 | 84 | |||
BC099852.1 | - | 2 | 84 | |||
BC099853.1 | - | 2 | 83 | |||
BC111805.1 | - | 2 | 77 | |||
BC141870.1 | - | 2 | 85 | 40038882 |