Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012044.2 | NP_036174.1 | 3 | 242 | ||
GenBank mRNA | AF112984.1 | - | 3 | 242 | ||
AF166098.1 | - | 3 | 238 | |||
AK148187.1 | - | 258 | ||||
BC027524.1 | - | 2 | 131 | 1533423 |