Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015758.2 | NP_056573.1 | 1 | 273 | ||
GenBank mRNA | AB601886.1 | - | 327 | |||
BC141057.1 | - | 1 | 312 | 9055669 |