Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018860.4 | NP_061348.1 | 4 | 244 | ||
GenBank mRNA | AK002503.1 | - | 4 | 190 | ||
AK007874.1 | - | 4 | 192 | |||
AK009548.1 | - | 4 | 191 | |||
AK010246.1 | - | 4 | 191 | |||
AK010317.1 | - | 4 | 191 | |||
AK010482.1 | - | 4 | 191 | |||
AK010509.1 | - | 4 | 191 | |||
AK010564.1 | - | 4 | 189 | |||
AK010583.1 | - | 4 | 191 | |||
AK010585.1 | - | 4 | 189 | |||
AK012252.1 | - | 4 | 189 | |||
AK012450.1 | - | 4 | 194 | |||
AK012476.1 | - | 4 | 190 | |||
AK012487.1 | - | 4 | 189 | |||
AK013324.1 | - | 4 | 196 | |||
AK019049.1 | - | 4 | 191 | |||
AK019178.1 | - | 4 | 186 | |||
AK028126.1 | - | 4 | 189 | |||
AK028231.1 | - | 4 | 192 | |||
AK028263.1 | - | 4 | 189 | |||
AK150502.1 | - | 4 | 191 | |||
AK151566.1 | - | 4 | 190 | |||
AK160588.1 | - | 4 | 191 | |||
AK168107.1 | - | 4 | 191 | |||
AK168485.1 | - | 4 | 189 | |||
AK168653.1 | - | 4 | 189 | |||
AK168719.1 | - | 4 | 189 | |||
AV478133.1 | - | 4 | 177 | |||
BC081450.1 | - | 3 | 260 | |||
BC083066.1 | - | 4 | 178 | |||
BC089572.1 | - | 4 | 188 | |||
BC089573.1 | - | 4 | 190 | |||
BC092010.1 | - | 4 | 179 | |||
BC099380.1 | - | 3 | 258 | |||
BC100592.1 | - | 4 | 190 | |||
BC145727.1 | - | 4 | 191 | 40131030 | ||
BC145729.1 | - | 4 | 191 | 40131032 | ||
BU937034.1 | - | 4 | 247 | |||
DQ113647.1 | - | 150 | ||||
U93862.1 | - | 3 | 165 |