Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slurp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020519.1 | NP_065265.1 | 2 | 150 | ||
GenBank mRNA | AK003904.1 | - | 2 | 148 | ||
AK029079.1 | - | 158 | ||||
BC111105.1 | - | 2 | 149 | 40052992 | ||
BC125244.1 | - | 2 | 149 | 40053004 |