Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024196.3 | NP_077158.1 | 8 | 1672 | ||
GenBank mRNA | AK036751.1 | - | 1588 | |||
AK076124.1 | - | 8 | 1673 | |||
AK082369.1 | - | 1822 | ||||
AK085702.1 | - | 1670 | ||||
AK158274.1 | - | 1669 | ||||
AK168985.1 | - | 1673 | ||||
AK195183.1 | - | 1 | 186 | |||
AK208244.1 | - | 1 | 185 | |||
BC034504.1 | - | 1654 | 4192736 | |||
BC086617.1 | - | 1658 |