Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026836.2 | NP_081112.1 | 5 | 1122 | ||
GenBank mRNA | AK004168.1 | - | 4 | 769 | ||
AK050689.1 | - | 5 | 1123 | |||
AK133851.1 | - | 6 | 2107 |