Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127487.1 | NP_001120959.1 | 20 | 3328 | ||
NM_001458.4 | NP_001449.3 | 20 | 3328 | |||
GenBank mRNA | AB371585.1 | - | 20 | 3348 | ||
AF089841.1 | - | 20 | 3168 | |||
AF146692.1 | - | 20 | 3331 | |||
AJ012737.1 | - | 20 | 3137 |