Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lce1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029667.2 | NP_083943.1 | 2 | 545 | ||
GenBank mRNA | AK010137.1 | - | 2 | 548 |