Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054037.2 | NP_473378.1 | 3 | 419 | ||
NM_170727.2 | NP_733923.1 | 4 | 524 | |||
GenBank mRNA | AF313456.1 | - | 3 | 404 | ||
AF313457.1 | - | 4 | 509 | |||
AK003988.1 | - | 3 | 406 | |||
BC086910.1 | - | 3 | 401 | 6759656 |