Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204201.1 | NP_001191130.1 | 8 | 1373 | ||
NM_001204202.1 | NP_001191131.1 | 7 | 1370 | |||
NM_001204203.1 | NP_001191132.1 | 8 | 1321 | |||
NM_001204233.1 | NP_001191162.1 | 6 | 1415 | |||
NM_009263.3 | NP_033289.2 | 7 | 1318 | |||
GenBank mRNA | AK149588.1 | - | 7 | 1309 | ||
AK150845.1 | - | 7 | 1309 | |||
AK151069.1 | - | 7 | 1279 | |||
AK151562.1 | - | 7 | 1309 | |||
AK153089.1 | - | 7 | 1308 | |||
AK154908.1 | - | 7 | 1309 | |||
AK160540.1 | - | 7 | 1312 | |||
AK165393.1 | - | 7 | 1309 | |||
AK165965.1 | - | 7 | 1360 | |||
BC057858.1 | - | 7 | 1291 | 5294167 | ||
J04806.1 | - | 7 | 1288 | |||
X13986.1 | - | 6 | 1233 | |||
X16151.1 | - | 6 | 1440 |