Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243093.1 | NP_001230022.1 | 15 | 2302 | ||
NM_001465.4 | NP_001456.3 | 15 | 2349 | |||
NM_199335.3 | NP_955367.1 | 14 | 2211 | |||
XM_006714464.2 | XP_006714527.1 | 15 | 2334 | |||
XM_006714465.2 | XP_006714528.1 | 16 | 2687 | |||
XM_006714466.2 | XP_006714529.1 | 15 | 2463 | |||
XM_011514008.2 | XP_011512310.1 | 17 | 2787 | |||
XM_011514009.1 | XP_011512311.1 | 17 | 2708 | |||
XM_011514010.1 | XP_011512312.1 | 16 | 2426 | |||
XM_011514011.2 | XP_011512313.1 | 14 | 2172 | |||
XM_017009316.1 | XP_016864805.1 | 14 | 2171 | |||
GenBank mRNA | AF001862.1 | - | 13 | 2115 | ||
AF198052.1 | - | 15 | 2186 | |||
AK290403.1 | - | 15 | 2344 | |||
AK297077.1 | - | 15 | 2302 | |||
BC015933.1 | - | 15 | 2278 | |||
BC117449.1 | - | 15 | 2248 | 40126000 | ||
BC143645.1 | - | 15 | 2248 | 9052159 | ||
BX647195.1 | - | 14 | 2196 | |||
BX647196.1 | - | 14 | 2197 | |||
CD701507.1 | - | 2 | 100 | |||
U93049.1 | - | 13 | 2078 |