Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap21-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028621.3 | NP_082897.2 | 1 | 444 | ||
GenBank mRNA | AK003736.2 | - | 1 | 475 | ||
BC145974.1 | - | 1 | 444 | |||
BC145976.1 | - | 1 | 444 | |||
BC147776.1 | - | 1 | 538 | 9087497 | ||
BC147786.1 | - | 1 | 538 | 9087505 |