Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010030.1 | NP_034160.1 | 2 | 135 | ||
GenBank mRNA | AJ011800.1 | - | 2 | 135 | ||
BC109129.1 | - | 2 | 123 | 40038941 | ||
BC109130.1 | - | 2 | 123 | 40038942 |