Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001487.3 | NP_001478.2 | 4 | 547 | ||
NR_037655.1 | 3 | 474 | ||||
NR_037656.1 | 4 | 810 | ||||
NR_037657.1 | 3 | 737 | ||||
GenBank mRNA | AK307069.1 | - | 3 | 818 | ||
BU601406.1 | - | 4 | 557 | |||
CN263524.1 | - | 3 | 522 | |||
D64007.1 | - | 4 | 543 | |||
S82447.1 | - | 4 | 500 |