Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012477.2 | NP_001012495.1 | 3 | 321 | ||
NM_013655.4 | NP_038683.1 | 3 | 321 | |||
NM_021704.3 | NP_068350.1 | 3 | 321 | |||
GenBank mRNA | AK045092.1 | - | 3 | 280 | ||
AK075596.1 | - | 3 | 280 | |||
AK157553.1 | - | 3 | 282 | |||
BB610070.1 | - | 3 | 277 | |||
BB840704.1 | - | 3 | 280 | |||
BC006640.1 | - | 3 | 269 | 3483088 | ||
BC020297.1 | - | 3 | 269 | |||
BC029335.1 | - | 3 | 269 | |||
BC046827.1 | - | 3 | 271 | |||
CT010389.1 | - | 3 | 200 | |||
D21072.1 | - | 3 | 258 | |||
D43804.1 | - | 3 | 278 | |||
D43805.1 | - | 3 | 278 | |||
L12029.1 | - | 3 | 281 | |||
L12030.1 | - | 3 | 276 | |||
S74318.1 | - | 3 | 266 |