Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021290.2 | NP_067265.1 | 2 | 456 | ||
GenBank mRNA | BC116948.1 | - | 2 | 390 | 40126267 | |
BC116950.1 | - | 2 | 390 | 40126269 | ||
BC144891.1 | - | 2 | 385 | 9053439 |