Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021431.2 | NP_067406.2 | 2 | 927 | ||
GenBank mRNA | AB041576.1 | - | 2 | 673 | ||
AK036077.1 | - | 2 | 258 | |||
AK053310.1 | - | 2 | 707 | |||
AY152853.1 | - | 2 | 673 | |||
BC071274.1 | - | 2 | 731 | 6331160 |