Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141927.1 | NP_001135399.1 | 3 | 422 | ||
NM_023129.5 | NP_075618.1 | 2 | 295 | |||
GenBank mRNA | AK002622.1 | - | 2 | 275 | ||
AK040718.1 | - | 2 | 266 | |||
AK052199.1 | - | 2 | 229 | |||
AK142708.1 | - | 2 | 250 | |||
BB658675.1 | - | 2 | 295 | |||
BC061097.1 | - | 2 | 288 | 6772294 | ||
CA459471.1 | - | 3 | 415 | |||
S46792.1 | - | 2 | 258 |