Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138751.2 | NP_620090.1 | 3 | 1993 | ||
GenBank mRNA | AB041540.1 | - | 3 | 1925 | ||
AK030761.1 | - | 3 | 1923 | |||
AK083563.1 | - | 1 | 1206 | |||
AK133790.1 | - | 3 | 1932 | |||
AK162265.1 | - | 1 | 612 | |||
AK162304.1 | - | 3 | 1930 | |||
BC019751.1 | - | 3 | 1904 | 4020610 |