Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bola2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175103.3 | NP_780312.1 | 2 | 174 | ||
GenBank mRNA | AK002662.1 | - | 2 | 177 | ||
AK003922.1 | - | 2 | 177 | |||
AK028165.1 | - | 2 | 188 | |||
AK140279.1 | - | 255 | ||||
BC045523.1 | - | 2 | 192 | 6397886 | ||
BC061197.1 | - | 2 | 202 | 6707992 |