Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178869.4 | NP_849200.2 | 12 | 1966 | ||
GenBank mRNA | AK031917.1 | - | 12 | 1937 | ||
AK140472.1 | - | 12 | 1937 | |||
AK168786.1 | - | 12 | 1937 | |||
AK169150.1 | - | 12 | 1953 | |||
AK179421.1 | - | 1 | 314 | |||
AK181337.1 | - | 1 | 306 | |||
AK182028.1 | - | 1 | 311 | |||
AK185779.1 | - | 1 | 276 | |||
AK216140.1 | - | 1 | 266 | |||
BC010510.1 | - | 12 | 1915 | 4191464 |