Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Soat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146064.1 | NP_666176.1 | 3, 4 | 340 | ||
GenBank mRNA | BC025931.1 | - | 3, 4 | 340 | 5099063 |