Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175653.2 | NP_783584.1 | 1 | 434 | ||
GenBank mRNA | BC120800.1 | - | 1 | 515 | 40129723 | |
BC120802.1 | - | 1 | 515 | 40129725 |