Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170488.1 | NP_001163959.1 | 7 | 1018 | ||
NM_176842.3 | NP_789812.1 | 6 | 910 | |||
GenBank mRNA | AK075592.1 | - | 1 | 82 | ||
AK217515.1 | - | 1 | 111 | |||
BC024858.1 | - | 6 | 873 | 5357332 | ||
BM948212.1 | - | 4 | 647 |