Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177583.4 | NP_808251.1 | 2 | 256 | ||
GenBank mRNA | AK028521.1 | - | 2 | 270 | ||
AK049828.1 | - | 2 | 257 | |||
AK154847.1 | - | 2 | 324 | |||
AK162585.1 | - | 2 | 238 | |||
BC079659.1 | - | 2 | 288 | 6853991 |