Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177613.2 | NP_808281.1 | 3, 4 | 581 | ||
GenBank mRNA | AK136078.1 | - | 557 | |||
BC039160.1 | - | 3, 4 | 581 | 4024472 | ||
BC094502.1 | - | 3, 4 | 397 | 5034261 |